Learn More
Aat Bioquest 5-FITC [Fluorescein-5-isothiocyanate] [CAS 3326-32-7]
SDP
Catalog No. 502979168
Encompass_Preferred
5-FITC and 6-FITC have very similar absorption and fluorescence spectra.
Catalog No. 50-297-9168 Supplier Aat Bioquest Supplier No. 122
5-FITC and 6-FITC have very similar absorption and fluorescence spectra.