Learn More
Cayman Chemical 5-fluoro MPP-PICA phenylpropionic acid metabolite, Purity: ≥98%, 10 mg
SDP
Catalog No. 502991515
Encompass_Preferred
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential metabolite of 5-fluoro MPP-PICA based on the published metabolism of 5-fluoro AMB-PICA; intended for research and forensic applications
Catalog No. 50-299-1515 Supplier Cayman Chemical Supplier No. 3127110 MG
An analytical reference standard that is structurally similar to known synthetic cannabinoids; a potential metabolite of 5-fluoro MPP-PICA based on the published metabolism of 5-fluoro AMB-PICA; intended for research and forensic applications