Learn More
Cayman Chemical 5fluoro nI 2naphthyl Iso 10mg
SDP
Catalog No. 502954120
Encompass_Preferred
A derivative of NNEI with a fluorine atom at the terminal carbon of the pentyl chain and the naphthyl group joined at the 2 rather than 1 position; intended for forensic and research applications
Catalog No. 50-295-4120 Supplier Cayman Chemical Supplier No. 1431810 MG
A derivative of NNEI with a fluorine atom at the terminal carbon of the pentyl chain and the naphthyl group joined at the 2 rather than 1 position; intended for forensic and research applications