Learn More
Cayman Chemical 5fluoro PB22 3hdroxquIno 10mg
SDP
Catalog No. 502969353
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the ester linkage inserted at the three, rather than at the eight, position of the quinoline moiety; intended for forensic and research purposes
Catalog No. 50-296-9353 Supplier Cayman Chemical Supplier No. 1451110 MG
Differs from 5-fluoro PB-22 by having the ester linkage inserted at the three, rather than at the eight, position of the quinoline moiety; intended for forensic and research purposes