Learn More
Cayman Chemical 5-fluoro PB-22 4-hydroxyisoquinoline isomer, Purity: ≥98%, CAS: 2365471-00-5, 10 mg
SDP
Catalog No. 502971011
Encompass_Preferred
Differs from 5-fluoro PB-22 by having an isoquinoline group attached at its four position in place of the quinoline group; intended for forensic and research purposes
Catalog No. 50-297-1011 Supplier Cayman Chemical Supplier No. 1454910 MG
Differs from 5-fluoro PB-22 by having an isoquinoline group attached at its four position in place of the quinoline group; intended for forensic and research purposes