Learn More
Cayman Chemical 5-fluoro PB-22 5-hydroxyisoquinoline isomer, Purity: ≥98%, CAS: 2365471-16-3, 10 mg
SDP
Catalog No. 502970060
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the quinoline group replaced with an isoquinoline group attached at its five position; intended for forensic and research purposes
Catalog No. 50-297-0060 Supplier Cayman Chemical Supplier No. 1455010 MG
Differs from 5-fluoro PB-22 by having the quinoline group replaced with an isoquinoline group attached at its five position; intended for forensic and research purposes