Learn More
Cayman Chemical 5fluoro PB22 5hdroxquIno 10mg
SDP
Catalog No. 502970666
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the ester linkage inserted at the fifth, rather than eighth, position of the quinoline moiety; intended for forensic and research purposes
Catalog No. 50-297-0666 Supplier Cayman Chemical Supplier No. 1451310 MG
Differs from 5-fluoro PB-22 by having the ester linkage inserted at the fifth, rather than eighth, position of the quinoline moiety; intended for forensic and research purposes