Learn More
Cayman Chemical 5-fluoro PB-22 6-hydroxyquinoline isomer, Purity: ≥98%, CAS: 2365471-03-8, 10 mg
SDP
Catalog No. 502970392
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the ester linkage inserted at the six, rather than eight, position of the quinoline moiety; intended for forensic and research purposes
Catalog No. 50-297-0392 Supplier Cayman Chemical Supplier No. 1451410 MG
Differs from 5-fluoro PB-22 by having the ester linkage inserted at the six, rather than eight, position of the quinoline moiety; intended for forensic and research purposes