Learn More
Cayman Chemical 5-fluoro PB-22 7-hydroxyisoquinoline isomer, Purity: ≥98%, CAS: 2365471-81-2, 10 mg
SDP
Catalog No. 502970668
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the quinoline group replaced with an isoquinoline group attached at its seven position; intended for forensic and research purposes
Catalog No. 50-297-0668 Supplier Cayman Chemical Supplier No. 1455210 MG
Differs from 5-fluoro PB-22 by having the quinoline group replaced with an isoquinoline group attached at its seven position; intended for forensic and research purposes