Learn More
Cayman Chemical 5-fluoro PB-22 7-hydroxyquinoline isomer, Purity: ≥98%, CAS: 1630756-70-5, 10 mg
SDP
Catalog No. 502971538
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the ester linkage inserted at the seven, rather than at the eight, position of the quinoline moiety; intended for forensic and research purposes.
Catalog No. 50-297-1538 Supplier Cayman Chemical Supplier No. 1451510 MG
Differs from 5-fluoro PB-22 by having the ester linkage inserted at the seven, rather than at the eight, position of the quinoline moiety; intended for forensic and research purposes.