Learn More
Cayman Chemical 5-fluoro PB-22 8-hydroxyisoquinoline isomer, Purity: ≥98%, CAS: 2365470-99-9, 5 mg
SDP
Catalog No. 502971080
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the quinoline group replaced with an isoquinoline group attached at its eight position; intended for forensic and research purposes
Catalog No. 50-297-1080 Supplier Cayman Chemical Supplier No. 145535 MG
Differs from 5-fluoro PB-22 by having the quinoline group replaced with an isoquinoline group attached at its eight position; intended for forensic and research purposes