Learn More
Cayman Chemical 5fluoro PB22 N 3fluoropen
SDP
Catalog No. 502971013
Encompass_Preferred
Differs from 5-fluoro PB-22 by having the fluorine atom at the 3 position, rather than the terminal 5 position, of the alkyl group; intended for forensic and research purposes
Catalog No. 50-297-1013 Supplier Cayman Chemical Supplier No. 14547500 UG
Differs from 5-fluoro PB-22 by having the fluorine atom at the 3 position, rather than the terminal 5 position, of the alkyl group; intended for forensic and research purposes