Learn More
Cayman Chemical 5Fluoropentyl3pyrIdInoyln 10mg
SDP
Catalog No. 502953997
Encompass_Preferred
A synthetic CB which shares the aminopentylindole structure of potent CBs like JWH 018
Catalog No. 50-295-3997 Supplier Cayman Chemical Supplier No. 1429510 MG
A synthetic CB which shares the aminopentylindole structure of potent CBs like JWH 018