Learn More
Air Science Usa Forensic Evidence Drying Cabinet, Freestanding, 60" Wide, 115V 60Hz
SDP
Catalog No. NC1626207
Encompass_Preferred
Safekeeper Series Forensic Evidence Drying Cabinet, Freestanding, 60" Wide, Single Chamber Version, Safekeeper series forensic evidence drying cabinets are designed to store, dry or otherwise process forensic evidence in the controlled environment of a tamper resistant compartment to maintain the chain of custody. The industry leading Multiplex filtration system, combined with professional design and construction offer convenience and protect the safety of personnel during use, maintenance and decontamination of the cabinets
Catalog No. NC1626207 Supplier Air Science Usa Supplier No. FDC008
Add to Cart
Add to Cart
Request a Quote
Safekeeper Series Forensic Evidence Drying Cabinet, Freestanding, 60" Wide, Single Chamber Version, Safekeeper series forensic evidence drying cabinets are designed to store, dry or otherwise process forensic evidence in the controlled environment of a tamper resistant compartment to maintain the chain of custody. The industry leading Multiplex filtration system, combined with professional design and construction offer convenience and protect the safety of personnel during use, maintenance and decontamination of the cabinets