Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, (1,3-Dimethyl-1H-pyrazole-5-yl)methanamine, 95%, [499770-63-7], D11377 125.172 C6H11N3

Catalog No. 5036563
Encompass_Preferred

Compound: (1,3-Dimethyl-1H-pyrazole-5-yl)methanamine

CAS#: 499770-63-7

MDL#: not listed

Background: A dimethylated pyrazole methanamine used as a building block in pharmaceutical research. The primary amine allows for amide coupling and further derivatization of the pyrazole heterocyclic scaffold.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-365-63 Supplier Frontier Specialty Chemicals Supplier No. D113775G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: (1,3-Dimethyl-1H-pyrazole-5-yl)methanamine

CAS#: 499770-63-7

MDL#: not listed

Background: A dimethylated pyrazole methanamine used as a building block in pharmaceutical research. The primary amine allows for amide coupling and further derivatization of the pyrazole heterocyclic scaffold.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.