Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 1,4-Dibromo-2,5-diiodobenzene, 99%, [63262-06-6], D2010 487.7 C6H2Br2I2

Catalog No. 50982168
Encompass_Preferred

Compound: 1,4-Dibromo-2,5-diiodobenzene

CAS#: 63262-06-6

MDL#: MFCD00082898

Background: A tetrahalogenated benzene bearing both bromine and iodine substituents, used as a precursor for constructing more complex chemical structures in organic synthesis and materials science. The differing reactivity of the bromine and iodine sites allows for sequential, selective cross-coupling.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-982-168 Supplier Frontier Specialty Chemicals Supplier No. D20105G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1,4-Dibromo-2,5-diiodobenzene

CAS#: 63262-06-6

MDL#: MFCD00082898

Background: A tetrahalogenated benzene bearing both bromine and iodine substituents, used as a precursor for constructing more complex chemical structures in organic synthesis and materials science. The differing reactivity of the bromine and iodine sites allows for sequential, selective cross-coupling.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.