Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 2,3-Dihydro-1-benzofuran-5-carboxaldehyde, 0.92, [55745-70-5], D12355 148.159 C9H8O2

Catalog No. 5036665
Encompass_Preferred

Compound: 2,3-Dihydro-1-benzofuran-5-carboxaldehyde

CAS#: 55745-70-5

MDL#: not listed

Background: A dihydrobenzofuran carbaldehyde used as a building block in pharmaceutical research. The aldehyde group provides a reactive handle for reductive amination and other condensation reactions in benzofuran-based scaffold synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-366-65 Supplier Frontier Specialty Chemicals Supplier No. D123555G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2,3-Dihydro-1-benzofuran-5-carboxaldehyde

CAS#: 55745-70-5

MDL#: not listed

Background: A dihydrobenzofuran carbaldehyde used as a building block in pharmaceutical research. The aldehyde group provides a reactive handle for reductive amination and other condensation reactions in benzofuran-based scaffold synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.