Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 2,5-Dimethoxyphenylboronic acid, 97%, [107099-99-0], D6276 181.98 C8H11BO4

Catalog No. 50982200
Encompass_Preferred

Compound: 2,5-Dimethoxyphenylboronic acid

CAS#: 107099-99-0

MDL#: MFCD01318181

Background: 2,5-Dimethoxyphenylboronic acid is an arylboronic acid used as a reactant in Suzuki-Miyaura and Negishi cross-coupling reactions. It is widely utilized in organic synthesis and medicinal chemistry, forming stable complexes with diols that make it useful in pharmaceutical and agrochemical development. It has also been studied for antileishmanial activity through inhibition of protein synthesis and cell division.

Catalog No. 50-982-200 Supplier Frontier Specialty Chemicals Supplier No. D62765G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2,5-Dimethoxyphenylboronic acid

CAS#: 107099-99-0

MDL#: MFCD01318181

Background: 2,5-Dimethoxyphenylboronic acid is an arylboronic acid used as a reactant in Suzuki-Miyaura and Negishi cross-coupling reactions. It is widely utilized in organic synthesis and medicinal chemistry, forming stable complexes with diols that make it useful in pharmaceutical and agrochemical development. It has also been studied for antileishmanial activity through inhibition of protein synthesis and cell division.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.