Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 2-Aminopyrimidine-5-boronic acid pinacol ester, 98%, [402960-38-7], A2103 221.07 C10H16BN3O2

Catalog No. 50981067
Encompass_Preferred

Compound: 2-Aminopyrimidine-5-boronic acid pinacol ester

CAS#: 402960-38-7

MDL#: MFCD06795685

Background: An amino-substituted pyrimidine boronic ester used as a coupling partner in Suzuki-Miyaura reactions. It has been used in the preparation of phosphoinositide-3-kinase (PI3K) inhibitors, supporting medicinal chemistry research into targeted therapies.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-981-067 Supplier Frontier Specialty Chemicals Supplier No. A21035G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Aminopyrimidine-5-boronic acid pinacol ester

CAS#: 402960-38-7

MDL#: MFCD06795685

Background: An amino-substituted pyrimidine boronic ester used as a coupling partner in Suzuki-Miyaura reactions. It has been used in the preparation of phosphoinositide-3-kinase (PI3K) inhibitors, supporting medicinal chemistry research into targeted therapies.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.