Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 2-Bromo-5-formylpyridine, >98%, [149806-06-4], B1609 186.01 C6H4BrNO

Catalog No. 50981297
Encompass_Preferred

Compound: 2-Bromo-5-formylpyridine

CAS#: 149806-06-4

MDL#: MFCD04115419

Background: 2-Bromo-5-formylpyridine is a highly functionalized pyridine building block whose bromine atom serves as a handle for palladium-catalyzed cross-coupling reactions such as Suzuki, Stille, and Heck couplings. Its aldehyde group readily undergoes nucleophilic addition, reduction, oxidation, or condensation to imines and oximes, and the compound is also used in Wittig reactions, providing versatile pathways for pharmaceutical and agrochemical synthesis.

Catalog No. 50-981-297 Supplier Frontier Specialty Chemicals Supplier No. B16095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Bromo-5-formylpyridine

CAS#: 149806-06-4

MDL#: MFCD04115419

Background: 2-Bromo-5-formylpyridine is a highly functionalized pyridine building block whose bromine atom serves as a handle for palladium-catalyzed cross-coupling reactions such as Suzuki, Stille, and Heck couplings. Its aldehyde group readily undergoes nucleophilic addition, reduction, oxidation, or condensation to imines and oximes, and the compound is also used in Wittig reactions, providing versatile pathways for pharmaceutical and agrochemical synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.