Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 2-Methoxypyridine-5-boronic acid, 98%, [163105-89-3], M5399 152.95 C6H8BNO3

Catalog No. 50982768
Encompass_Preferred

Compound: 2-Methoxypyridine-5-boronic acid

CAS#: 163105-89-3

MDL#: MFCD02093044

Background: A methoxy-substituted pyridine boronic acid used as a coupling partner in Suzuki-Miyaura cross-coupling reactions. It is widely utilized in organic synthesis and medicinal chemistry for forming carbon-carbon bonds in the construction of substituted pyridine scaffolds.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-982-768 Supplier Frontier Specialty Chemicals Supplier No. M53995G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Methoxypyridine-5-boronic acid

CAS#: 163105-89-3

MDL#: MFCD02093044

Background: A methoxy-substituted pyridine boronic acid used as a coupling partner in Suzuki-Miyaura cross-coupling reactions. It is widely utilized in organic synthesis and medicinal chemistry for forming carbon-carbon bonds in the construction of substituted pyridine scaffolds.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.