Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 2-Methylprop-1-enylboronic acid pinacol ester, 98%, [126689-00-7], M11191 182.068 C10H19BO2

Catalog No. 5038217
Encompass_Preferred

Compound: 2-Methylprop-1-enylboronic acid pinacol ester

CAS#: 126689-00-7

MDL#: not listed

Background: A branched alkenylboronic ester used as a Suzuki-Miyaura coupling partner in organic synthesis. It provides a convenient means to introduce an isobutenyl group into pharmaceutical and materials science building blocks.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-382-17 Supplier Frontier Specialty Chemicals Supplier No. M111915G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Methylprop-1-enylboronic acid pinacol ester

CAS#: 126689-00-7

MDL#: not listed

Background: A branched alkenylboronic ester used as a Suzuki-Miyaura coupling partner in organic synthesis. It provides a convenient means to introduce an isobutenyl group into pharmaceutical and materials science building blocks.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.