Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, (2-Methylpropyl)boronic acid, 97%, [84110-40-7], I7111 101.94 C4H11BO2

Catalog No. 50982613
Encompass_Preferred

Compound: (2-Methylpropyl)boronic acid

CAS#: 84110-40-7

MDL#: MFCD00134156

Background: (2-Methylpropyl)boronic acid, also known as isobutylboronic acid, has been used to synthesize potent and selective dipeptidyl boronic acid proteasome inhibitors studied as therapeutics for cancer and inflammatory diseases, and in the preparation of isocoumarin analogs as gamma-secretase cleavage inhibitors. It is also used broadly in Suzuki-Miyaura reactions for the synthesis of complex pharmaceutical and agrochemical molecules.

Catalog No. 50-982-613 Supplier Frontier Specialty Chemicals Supplier No. I71115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: (2-Methylpropyl)boronic acid

CAS#: 84110-40-7

MDL#: MFCD00134156

Background: (2-Methylpropyl)boronic acid, also known as isobutylboronic acid, has been used to synthesize potent and selective dipeptidyl boronic acid proteasome inhibitors studied as therapeutics for cancer and inflammatory diseases, and in the preparation of isocoumarin analogs as gamma-secretase cleavage inhibitors. It is also used broadly in Suzuki-Miyaura reactions for the synthesis of complex pharmaceutical and agrochemical molecules.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.