Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 3-Acrylamidophenylboronic acid, 98%, [99349-68-5], A10094 1999% C9H10BNO3

Catalog No. 50981010
Encompass_Preferred

Compound: 3-Acrylamidophenylboronic acid

CAS#: 99349-68-5

MDL#: MFCD09025755

Background: A boronic acid monomer bearing an acrylamide group, known as AAPBA, that forms reversible covalent bonds with diols and sugars. It is used to synthesize glucose-sensitive boronic acid-based polymers and copolymers for biosensors, drug delivery systems, and glucose-monitoring applications.

Storage: Store at -20°C, under dry conditions.

Catalog No. 50-981-010 Supplier Frontier Specialty Chemicals Supplier No. A100945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Acrylamidophenylboronic acid

CAS#: 99349-68-5

MDL#: MFCD09025755

Background: A boronic acid monomer bearing an acrylamide group, known as AAPBA, that forms reversible covalent bonds with diols and sugars. It is used to synthesize glucose-sensitive boronic acid-based polymers and copolymers for biosensors, drug delivery systems, and glucose-monitoring applications.

Storage: Store at -20°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.