Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 3-Bromo-4-formylthiophene, 98%, [18791-78-1], B7762 191.05 C5H3BrOS

Catalog No. 50981729
Encompass_Preferred

Compound: 3-Bromo-4-formylthiophene

CAS#: 18791-78-1

MDL#: MFCD01318960

Background: A brominated thiophene aldehyde used in organic synthesis, including dehalogenation and hydrogenation studies. It also serves as a building block for near-infrared-absorbing polymer dyes used in materials science applications such as organic solar cells.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-729 Supplier Frontier Specialty Chemicals Supplier No. B77625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Bromo-4-formylthiophene

CAS#: 18791-78-1

MDL#: MFCD01318960

Background: A brominated thiophene aldehyde used in organic synthesis, including dehalogenation and hydrogenation studies. It also serves as a building block for near-infrared-absorbing polymer dyes used in materials science applications such as organic solar cells.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.