Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 3-Chloropyridine-4-boronic acid hydrate, 96%, [1256355-22-2], C1827 175.378 C5H7BClNO3

Catalog No. 50981885
Encompass_Preferred

Compound: 3-Chloropyridine-4-boronic acid hydrate

CAS#: 1256355-22-2

MDL#: MFCD03453058

Background: 3-Chloropyridine-4-boronic acid hydrate is offered for experimental and research use as a synthetic building block in organic chemistry. Like related chloropyridine boronic acid derivatives, it is used in Suzuki-Miyaura cross-coupling reactions, which are essential for forming carbon-carbon bonds in pharmaceutical and agrochemical development.

Catalog No. 50-981-885 Supplier Frontier Specialty Chemicals Supplier No. C18275G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Chloropyridine-4-boronic acid hydrate

CAS#: 1256355-22-2

MDL#: MFCD03453058

Background: 3-Chloropyridine-4-boronic acid hydrate is offered for experimental and research use as a synthetic building block in organic chemistry. Like related chloropyridine boronic acid derivatives, it is used in Suzuki-Miyaura cross-coupling reactions, which are essential for forming carbon-carbon bonds in pharmaceutical and agrochemical development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.