Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 3-Chloropyridine-5-boronic acid, 97%, [872041-85-5], C10087 157.36 C5H5BClNO2

Catalog No. 50981769
Encompass_Preferred

Compound: 3-Chloropyridine-5-boronic acid

CAS#: 872041-85-5

MDL#: MFCD06798243

Background: A chlorinated pyridine boronic acid used in Suzuki-Miyaura cross-coupling reactions. It has been used in the preparation of the PET radioligand [11C]MK-1064 for orexin-2 receptor imaging, the orexin 2 receptor antagonist MK-1064, and ATAD2 bromodomain inhibitors in pharmaceutical research.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-981-769 Supplier Frontier Specialty Chemicals Supplier No. C100875G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Chloropyridine-5-boronic acid

CAS#: 872041-85-5

MDL#: MFCD06798243

Background: A chlorinated pyridine boronic acid used in Suzuki-Miyaura cross-coupling reactions. It has been used in the preparation of the PET radioligand [11C]MK-1064 for orexin-2 receptor imaging, the orexin 2 receptor antagonist MK-1064, and ATAD2 bromodomain inhibitors in pharmaceutical research.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.