Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 3-(N,N-Dimethylcarbamoyl)phenylboronic acid, 98%, [373384-14-6], D4088 193 C9H12BNO3

Catalog No. 50982179
Encompass_Preferred

Compound: 3-(N,N-Dimethylcarbamoyl)phenylboronic acid

CAS#: 373384-14-6

MDL#: MFCD03412066

Background: 3-(N,N-Dimethylcarbamoyl)phenylboronic acid is used as a building block in pharmaceutical development, including the synthesis of biologically active molecules such as potential anti-cancer agents. It is also utilized in agrochemical synthesis and materials science, where its ability to participate in Suzuki-Miyaura cross-coupling reactions supports the development of novel polymers and nanomaterials.

Catalog No. 50-982-179 Supplier Frontier Specialty Chemicals Supplier No. D40885G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-(N,N-Dimethylcarbamoyl)phenylboronic acid

CAS#: 373384-14-6

MDL#: MFCD03412066

Background: 3-(N,N-Dimethylcarbamoyl)phenylboronic acid is used as a building block in pharmaceutical development, including the synthesis of biologically active molecules such as potential anti-cancer agents. It is also utilized in agrochemical synthesis and materials science, where its ability to participate in Suzuki-Miyaura cross-coupling reactions supports the development of novel polymers and nanomaterials.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.