Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 3-Fluoro-5-methoxyphenylboronic acid, 98%, [609807-25-2], F2100 169.95 C7H8BFO3

Catalog No. 50982478
Encompass_Preferred

Compound: 3-Fluoro-5-methoxyphenylboronic acid

CAS#: 609807-25-2

MDL#: MFCD07366780

Background: 3-Fluoro-5-methoxyphenylboronic acid is used for proteomics research and forms stable complexes with diols, making it an essential reagent in Suzuki-Miyaura cross-coupling reactions. It is used in the development of pharmaceuticals, agrochemicals, and advanced materials, as well as in biosensing and diagnostic applications due to its selective binding to specific biomolecules.

Catalog No. 50-982-478 Supplier Frontier Specialty Chemicals Supplier No. F21005G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Fluoro-5-methoxyphenylboronic acid

CAS#: 609807-25-2

MDL#: MFCD07366780

Background: 3-Fluoro-5-methoxyphenylboronic acid is used for proteomics research and forms stable complexes with diols, making it an essential reagent in Suzuki-Miyaura cross-coupling reactions. It is used in the development of pharmaceuticals, agrochemicals, and advanced materials, as well as in biosensing and diagnostic applications due to its selective binding to specific biomolecules.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.