Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 4-(1-Naphthyl)phenyl boronic acid, 98%, [870774-25-7], N10084 248.084 C16H13BO2

Catalog No. 5038510
Encompass_Preferred

Compound: 4-(1-Naphthyl)phenyl boronic acid

CAS#: 870774-25-7

MDL#: not listed

Background: A naphthyl-substituted phenylboronic acid used as a Suzuki-Miyaura coupling partner in materials science and pharmaceutical research. Naphthyl-biaryl building blocks of this type are commonly used in organic electronic and photoluminescent materials.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-385-10 Supplier Frontier Specialty Chemicals Supplier No. N100845G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-(1-Naphthyl)phenyl boronic acid

CAS#: 870774-25-7

MDL#: not listed

Background: A naphthyl-substituted phenylboronic acid used as a Suzuki-Miyaura coupling partner in materials science and pharmaceutical research. Naphthyl-biaryl building blocks of this type are commonly used in organic electronic and photoluminescent materials.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.