Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 4-Bromo-1-methyl-1H-pyrazole-5-carboxaldehyde, 95%, [473528-88-0], B12945 189.01 C5H5BrN2O

Catalog No. 5035937
Encompass_Preferred

Compound: 4-Bromo-1-methyl-1H-pyrazole-5-carboxaldehyde

CAS#: 473528-88-0

MDL#: not listed

Background: A brominated, N-methylated pyrazole carbaldehyde used as a building block in pharmaceutical research. The bromine substituent offers a reactive site for cross-coupling, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-359-37 Supplier Frontier Specialty Chemicals Supplier No. B129455G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Bromo-1-methyl-1H-pyrazole-5-carboxaldehyde

CAS#: 473528-88-0

MDL#: not listed

Background: A brominated, N-methylated pyrazole carbaldehyde used as a building block in pharmaceutical research. The bromine substituent offers a reactive site for cross-coupling, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.