Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 4-Carbamoylphenylboronic acid, 98%, [123088-59-5], B4488 164.95 C7H8BNO3

Catalog No. 50981651
Encompass_Preferred

Compound: 4-Carbamoylphenylboronic acid

CAS#: 123088-59-5

MDL#: MFCD03411940

Background: 4-Carbamoylphenylboronic acid is an arylboronic acid bearing a primary amide group. It is used as a reactant in Suzuki-Miyaura, Sonogashira, and Buchwald-Hartwig cross-coupling reactions to build substituted aromatic scaffolds. It has also served in the synthesis of biologically active molecules, including protein inhibitors and receptor antagonists.

Catalog No. 50-981-651 Supplier Frontier Specialty Chemicals Supplier No. B44885G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Carbamoylphenylboronic acid

CAS#: 123088-59-5

MDL#: MFCD03411940

Background: 4-Carbamoylphenylboronic acid is an arylboronic acid bearing a primary amide group. It is used as a reactant in Suzuki-Miyaura, Sonogashira, and Buchwald-Hartwig cross-coupling reactions to build substituted aromatic scaffolds. It has also served in the synthesis of biologically active molecules, including protein inhibitors and receptor antagonists.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.