Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 4-Chloro-2-(hydroxymethyl)pyridine, 98%, [63071-10-3], C10194 143.571 C6H6ClNO

Catalog No. 50981778
Encompass_Preferred

Compound: 4-Chloro-2-(hydroxymethyl)pyridine

CAS#: 63071-10-3

MDL#: MFCD07437885

Background: A chlorinated hydroxymethylpyridine used as an important intermediate in the synthesis of pharmaceuticals, particularly drugs targeting neurological disorders, as well as agrochemicals and other fine chemicals.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-981-778 Supplier Frontier Specialty Chemicals Supplier No. C101945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Chloro-2-(hydroxymethyl)pyridine

CAS#: 63071-10-3

MDL#: MFCD07437885

Background: A chlorinated hydroxymethylpyridine used as an important intermediate in the synthesis of pharmaceuticals, particularly drugs targeting neurological disorders, as well as agrochemicals and other fine chemicals.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.