Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 4-Fluoro-3-formylphenylboronic acid, 98%, [374538-01-9], F10861 167.93 C7H6BFO3

Catalog No. 5036898
Encompass_Preferred

Compound: 4-Fluoro-3-formylphenylboronic acid

CAS#: 374538-01-9

MDL#: not listed

Background: A fluorinated, formyl-substituted phenylboronic acid used as a bifunctional building block in pharmaceutical research. The aldehyde group provides a reactive handle for condensation reactions alongside Suzuki-Miyaura coupling chemistry.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-368-98 Supplier Frontier Specialty Chemicals Supplier No. F108615G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-Fluoro-3-formylphenylboronic acid

CAS#: 374538-01-9

MDL#: not listed

Background: A fluorinated, formyl-substituted phenylboronic acid used as a bifunctional building block in pharmaceutical research. The aldehyde group provides a reactive handle for condensation reactions alongside Suzuki-Miyaura coupling chemistry.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.