Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 4-(Methoxycarbonyl)phenylboronic acid, 97%, [99768-12-4], M3563 179.97 C8H9BO4

Catalog No. 50982758
Encompass_Preferred

Compound: 4-(Methoxycarbonyl)phenylboronic acid

CAS#: 99768-12-4

MDL#: MFCD01632203

Background: 4-(Methoxycarbonyl)phenylboronic acid is an arylboronic acid used as a key intermediate in pharmaceutical synthesis. It participates in Suzuki cross-coupling reactions to form carbon-carbon bonds for the construction of biaryl scaffolds. It has been used in the synthesis of HIV-1 maturation inhibitors and other drug candidates.

Catalog No. 50-982-758 Supplier Frontier Specialty Chemicals Supplier No. M35635G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-(Methoxycarbonyl)phenylboronic acid

CAS#: 99768-12-4

MDL#: MFCD01632203

Background: 4-(Methoxycarbonyl)phenylboronic acid is an arylboronic acid used as a key intermediate in pharmaceutical synthesis. It participates in Suzuki cross-coupling reactions to form carbon-carbon bonds for the construction of biaryl scaffolds. It has been used in the synthesis of HIV-1 maturation inhibitors and other drug candidates.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.