Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 4-(N-Boc-aminomethyl)phenylboronic acid, 97%, [489446-42-6], A1231 251.09 C12H18BNO4

Catalog No. 50981028
Encompass_Preferred

Compound: 4-(N-Boc-aminomethyl)phenylboronic acid

CAS#: 489446-42-6

MDL#: MFCD04115637

Background: 4-(N-Boc-aminomethyl)phenylboronic acid is a Boc-protected phenylboronic acid used for proteomics research and as a key building block in pharmaceutical and agrochemical synthesis. Its boronic acid functionality supports Suzuki-Miyaura cross-coupling reactions, while the protected amino group enables selective manipulation of specific sites within a molecule during multi-step synthesis.

Catalog No. 50-981-028 Supplier Frontier Specialty Chemicals Supplier No. A12315G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-(N-Boc-aminomethyl)phenylboronic acid

CAS#: 489446-42-6

MDL#: MFCD04115637

Background: 4-(N-Boc-aminomethyl)phenylboronic acid is a Boc-protected phenylboronic acid used for proteomics research and as a key building block in pharmaceutical and agrochemical synthesis. Its boronic acid functionality supports Suzuki-Miyaura cross-coupling reactions, while the protected amino group enables selective manipulation of specific sites within a molecule during multi-step synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.