Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 4-(N-Methylaminocarbonyl)phenylboronic acid, 98%, [121177-82-0], M10870 178.981 C8H10BNO3

Catalog No. 5038165
Encompass_Preferred

Compound: 4-(N-Methylaminocarbonyl)phenylboronic acid

CAS#: 121177-82-0

MDL#: not listed

Background: A secondary amide-substituted phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical research. The amide N-H offers a site for hydrogen bonding, useful for tuning the physicochemical properties of downstream coupled products.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-381-65 Supplier Frontier Specialty Chemicals Supplier No. M108705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 4-(N-Methylaminocarbonyl)phenylboronic acid

CAS#: 121177-82-0

MDL#: not listed

Background: A secondary amide-substituted phenylboronic acid used as a Suzuki-Miyaura coupling partner in pharmaceutical research. The amide N-H offers a site for hydrogen bonding, useful for tuning the physicochemical properties of downstream coupled products.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.