Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 5-Bromo-1H-pyrazolo[3,4-c]pyridine, 98%, [929617-35-6], B1216 193.43 C4H2BrClN2

Catalog No. 50981205
Encompass_Preferred

Compound: 5-Bromo-1H-pyrazolo[3,4-c]pyridine

CAS#: 929617-35-6

MDL#: MFCD00483232

Background: 5-Bromo-1H-pyrazolo[3,4-c]pyridine is a heterocyclic research chemical and synthetic building block used in pharmaceutical research, particularly in the synthesis of pyrazolopyridine derivatives investigated for potential therapeutic applications.

Catalog No. 50-981-205 Supplier Frontier Specialty Chemicals Supplier No. B12165G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromo-1H-pyrazolo[3,4-c]pyridine

CAS#: 929617-35-6

MDL#: MFCD00483232

Background: 5-Bromo-1H-pyrazolo[3,4-c]pyridine is a heterocyclic research chemical and synthetic building block used in pharmaceutical research, particularly in the synthesis of pyrazolopyridine derivatives investigated for potential therapeutic applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.