Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 5-Bromo-2-fluoropyrimidine, >98%, [62802-38-4], B10152 176.98 C4H2BrFN2

Catalog No. 50981101
Encompass_Preferred

Compound: 5-Bromo-2-fluoropyrimidine

CAS#: 62802-38-4

MDL#: MFCD07787364

Background: A halogenated pyrimidine used as a biochemical reagent in proteomics research. The fluoride group supports nucleophilic aromatic substitution while the bromide group enables palladium-catalyzed carbon-carbon coupling, making this compound useful for constructing electron-pulling push-pull molecules.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-101 Supplier Frontier Specialty Chemicals Supplier No. B101525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromo-2-fluoropyrimidine

CAS#: 62802-38-4

MDL#: MFCD07787364

Background: A halogenated pyrimidine used as a biochemical reagent in proteomics research. The fluoride group supports nucleophilic aromatic substitution while the bromide group enables palladium-catalyzed carbon-carbon coupling, making this compound useful for constructing electron-pulling push-pull molecules.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.