Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 5-Bromo-2-formylpyridine, 98%, [31181-90-5], B1610 186.01 C6H4BrNO

Catalog No. 50981300
Encompass_Preferred

Compound: 5-Bromo-2-formylpyridine

CAS#: 31181-90-5

MDL#: MFCD04112535

Background: 5-Bromo-2-formylpyridine is used as an organic chemical synthesis intermediate for producing a wide range of compounds, including pharmaceuticals and agrochemicals. Its bromine and aldehyde functional groups enable nucleophilic addition and coupling reactions, making it a valuable building block for exploring new synthetic pathways in medicinal chemistry and materials science.

Catalog No. 50-981-300 Supplier Frontier Specialty Chemicals Supplier No. B16105G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromo-2-formylpyridine

CAS#: 31181-90-5

MDL#: MFCD04112535

Background: 5-Bromo-2-formylpyridine is used as an organic chemical synthesis intermediate for producing a wide range of compounds, including pharmaceuticals and agrochemicals. Its bromine and aldehyde functional groups enable nucleophilic addition and coupling reactions, making it a valuable building block for exploring new synthetic pathways in medicinal chemistry and materials science.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.