Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 5-Bromo-2-hydrazinopyridine, 98%, [77992-44-0], B1870 188.03 C5H6BrN3

Catalog No. 50981513
Encompass_Preferred

Compound: 5-Bromo-2-hydrazinopyridine

CAS#: 77992-44-0

MDL#: MFCD00513424

Background: A brominated hydrazinopyridine used as an intermediate in pharmaceutical manufacturing for the synthesis of heterocyclic compounds. Its bromine and hydrazine functional groups make it valuable for complex organic synthesis, including the construction of fused heterocyclic ring systems.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-981-513 Supplier Frontier Specialty Chemicals Supplier No. B18705G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Bromo-2-hydrazinopyridine

CAS#: 77992-44-0

MDL#: MFCD00513424

Background: A brominated hydrazinopyridine used as an intermediate in pharmaceutical manufacturing for the synthesis of heterocyclic compounds. Its bromine and hydrazine functional groups make it valuable for complex organic synthesis, including the construction of fused heterocyclic ring systems.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.