Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 5-Formyl-1-methyl-1H-pyrazole, 98%, [27258-33-9], F2044 110.12 C5H6N2O

Catalog No. 50982472
Encompass_Preferred

Compound: 5-Formyl-1-methyl-1H-pyrazole

CAS#: 27258-33-9

MDL#: MFCD03419801

Background: A methylated pyrazole carbaldehyde used as a building block in the synthesis of pharmaceuticals and agrochemicals. It has been used as an intermediate in the development of pyrazole-based compounds studied for anti-inflammatory and anti-cancer properties.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-982-472 Supplier Frontier Specialty Chemicals Supplier No. F20445G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Formyl-1-methyl-1H-pyrazole

CAS#: 27258-33-9

MDL#: MFCD03419801

Background: A methylated pyrazole carbaldehyde used as a building block in the synthesis of pharmaceuticals and agrochemicals. It has been used as an intermediate in the development of pyrazole-based compounds studied for anti-inflammatory and anti-cancer properties.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.