Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, tert-Butyl 5-formylpyridin-2-ylcarbamate, 95%, [199296-40-7], F10425 222.24 C11H14N2O3

Catalog No. 5036861
Encompass_Preferred

Compound: tert-Butyl 5-formylpyridin-2-ylcarbamate

CAS#: 199296-40-7

MDL#: MFCD08064228

Background: A Boc-protected aminopyridine carbaldehyde used as a building block in pharmaceutical research. The aldehyde group supports reductive amination chemistry, while the carbamate can later be deprotected to reveal a free amine.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-368-61 Supplier Frontier Specialty Chemicals Supplier No. F104255G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: tert-Butyl 5-formylpyridin-2-ylcarbamate

CAS#: 199296-40-7

MDL#: MFCD08064228

Background: A Boc-protected aminopyridine carbaldehyde used as a building block in pharmaceutical research. The aldehyde group supports reductive amination chemistry, while the carbamate can later be deprotected to reveal a free amine.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.