Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 5-Formylpyrimidine, >97%, [10070-92-5], P1680 108.1 C5H4N2O

Catalog No. 50982934
Encompass_Preferred

Compound: 5-Formylpyrimidine

CAS#: 10070-92-5

MDL#: MFCD03426065

Background: A pyrimidine carbaldehyde widely utilized as a key intermediate in the synthesis of pharmaceuticals and agrochemicals. It is used as a building block in the preparation of biologically active molecules, including in reactions to form chiral pyrimidyl alkanols.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-982-934 Supplier Frontier Specialty Chemicals Supplier No. P16805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Formylpyrimidine

CAS#: 10070-92-5

MDL#: MFCD03426065

Background: A pyrimidine carbaldehyde widely utilized as a key intermediate in the synthesis of pharmaceuticals and agrochemicals. It is used as a building block in the preparation of biologically active molecules, including in reactions to form chiral pyrimidyl alkanols.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.