Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 5-Formylthiazole, 97%, [1003-32-3], F11307 113.138 C4H3NOS

Catalog No. 5036956
Encompass_Preferred

Compound: 5-Formylthiazole

CAS#: 1003-32-3

MDL#: not listed

Background: A thiazole carbaldehyde used as a building block in pharmaceutical research. The aldehyde group provides a reactive handle for reductive amination and other condensation reactions in thiazole-based scaffold synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-369-56 Supplier Frontier Specialty Chemicals Supplier No. F113075G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Formylthiazole

CAS#: 1003-32-3

MDL#: not listed

Background: A thiazole carbaldehyde used as a building block in pharmaceutical research. The aldehyde group provides a reactive handle for reductive amination and other condensation reactions in thiazole-based scaffold synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.