Learn More
MSC PRO-SAFE Drum Liner, 5 Gal, 15 mil, LDPE, Antistatic, 100/Pk.
Catalog No. 502886273
Encompass
This PRO-SAFE drum liner is designed for 5-gallon drums, featuring an antistatic LDPE material with a 15 mil thickness, ensuring safe containment and protection.
  • Rigid smooth liner
  • Compatible with 5 gal drums
  • Features 15 mil thickness
  • Made from LDPE material
  • Provides antistatic properties
  • Helps save paint buckets in plants
Catalog No. 50-288-6273 Supplier MSC Supplier No. 07509714
This PRO-SAFE drum liner is designed for 5-gallon drums, featuring an antistatic LDPE material with a 15 mil thickness, ensuring safe containment and protection.
  • Rigid smooth liner
  • Compatible with 5 gal drums
  • Features 15 mil thickness
  • Made from LDPE material
  • Provides antistatic properties
  • Helps save paint buckets in plants