Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Bis(pinacolato)diboron, 99%, [73183-34-3], D6878 253.94 C12H24B2O4

Catalog No. 50982217
Encompass_Preferred

Compound: Bis(pinacolato)diboron

CAS#: 73183-34-3

MDL#: MFCD00799570

Background: Bis(pinacolato)diboron is a versatile diboron reagent used with iridium or other transition-metal catalysts for the C-H borylation of arenes, heteroarenes, and benzylic positions of alkylarenes, furnishing aryl-, heteroaryl-, and benzylboron compounds. It reacts with a range of functional groups and scaffolds under metal-catalyzed conditions, including polymers and terminal alkenes, to generate diols. It is widely used to prepare boronate esters for Suzuki-Miyaura coupling and other palladium-catalyzed transformations.

Storage: Store at -20°C, under dry conditions.

Catalog No. 50-982-217 Supplier Frontier Specialty Chemicals Supplier No. D68785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Bis(pinacolato)diboron

CAS#: 73183-34-3

MDL#: MFCD00799570

Background: Bis(pinacolato)diboron is a versatile diboron reagent used with iridium or other transition-metal catalysts for the C-H borylation of arenes, heteroarenes, and benzylic positions of alkylarenes, furnishing aryl-, heteroaryl-, and benzylboron compounds. It reacts with a range of functional groups and scaffolds under metal-catalyzed conditions, including polymers and terminal alkenes, to generate diols. It is widely used to prepare boronate esters for Suzuki-Miyaura coupling and other palladium-catalyzed transformations.

Storage: Store at -20°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.