Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Cyclohexylboronic acid, 99%, [4441-56-9], C4722 127.98 C6H13BO2

Catalog No. 50981981
Encompass_Preferred

Compound: Cyclohexylboronic acid

CAS#: 4441-56-9

MDL#: MFCD01311824

Background: Cyclohexylboronic acid is an alkylboronic acid used as a reagent in Suzuki cross-coupling reactions to form carbon-carbon bonds. It supports the synthesis of biaryl and alkyl-aryl compounds relevant to pharmaceuticals, agrochemicals, and materials science. It is also used in the arylation and alkylation of heterocyclic scaffolds such as diphenylisoxazole.

Catalog No. 50-981-981 Supplier Frontier Specialty Chemicals Supplier No. C47225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Cyclohexylboronic acid

CAS#: 4441-56-9

MDL#: MFCD01311824

Background: Cyclohexylboronic acid is an alkylboronic acid used as a reagent in Suzuki cross-coupling reactions to form carbon-carbon bonds. It supports the synthesis of biaryl and alkyl-aryl compounds relevant to pharmaceuticals, agrochemicals, and materials science. It is also used in the arylation and alkylation of heterocyclic scaffolds such as diphenylisoxazole.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.