Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Cyclopropylboronic acid pinacol ester, 98%, [126689-01-8], C1471 168.05 C9H17BO2

Catalog No. 50981844
Encompass_Preferred

Compound: Cyclopropylboronic acid pinacol ester

CAS#: 126689-01-8

MDL#: MFCD05663847

Background: A cyclopropyl boronic ester used as a stable coupling partner in Suzuki-Miyaura cross-coupling reactions to install cyclopropyl groups onto aromatic and heteroaromatic scaffolds. It has been applied in the synthesis of FLAP inhibitors and other pharmaceutically relevant compounds.

Storage: Store at 2-8°C, under dry conditions.

Catalog No. 50-981-844 Supplier Frontier Specialty Chemicals Supplier No. C14715G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Cyclopropylboronic acid pinacol ester

CAS#: 126689-01-8

MDL#: MFCD05663847

Background: A cyclopropyl boronic ester used as a stable coupling partner in Suzuki-Miyaura cross-coupling reactions to install cyclopropyl groups onto aromatic and heteroaromatic scaffolds. It has been applied in the synthesis of FLAP inhibitors and other pharmaceutically relevant compounds.

Storage: Store at 2-8°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.