Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, Dibenzofuran-4-boronic acid, 97%, [100124-06-9], D5438 212.01 C12H9BO3

Catalog No. 50982184
Encompass_Preferred

Compound: Dibenzofuran-4-boronic acid

CAS#: 100124-06-9

MDL#: MFCD00092336

Background: Dibenzofuran-4-boronic acid is a heteroaryl boronic acid used as a coupling partner in Suzuki-Miyaura cross-coupling reactions to form biaryl and heterobiaryl structures. It has been used to prepare glycosensors, as its fluorescence changes upon binding sugars under near-physiological conditions. It also serves as an intermediate in pharmaceutical, agrochemical, and OLED material synthesis.

Catalog No. 50-982-184 Supplier Frontier Specialty Chemicals Supplier No. D54385G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Dibenzofuran-4-boronic acid

CAS#: 100124-06-9

MDL#: MFCD00092336

Background: Dibenzofuran-4-boronic acid is a heteroaryl boronic acid used as a coupling partner in Suzuki-Miyaura cross-coupling reactions to form biaryl and heterobiaryl structures. It has been used to prepare glycosensors, as its fluorescence changes upon binding sugars under near-physiological conditions. It also serves as an intermediate in pharmaceutical, agrochemical, and OLED material synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.